Skip to Content
Merck
Bis(triphenylphosphine)palladium(II) diacetate 98%

Bis(triphenylphosphine)palladium(II) diacetate

Bis(triphenylphosphine)palladium(II) diacetate 98%
Synonym(s):
Bis(acetato)bis(triphenylphosphine)palladium(II), Palladium(II)bis(triphenylphosphine) diacetate, Pd(OAc)2(PPh3)2
Linear Formula:
[(C6H5)3P]2Pd(CH3COO)2
CAS Number:
Molecular Weight:
749.08
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing