
Bis(tert-butylimino)bis(dimethylamino)tungsten(VI)

Linear Formula:
((CH3)3CN)2W(N(CH3)2)2
CAS Number:
Molecular Weight:
414.23
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2