
Bisphenol AP

Synonym(s):
4,4′-(1-Phenylethylidene)bisphenol, Bisphenol AP
Linear Formula:
CH3C(C6H5)(C6H4OH)2
CAS Number:
Molecular Weight:
290.36
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing