Skip to Content
Merck
Bismarck Brown Y (G)

Bismarck Brown Y (G)

Bismarck Brown Y (G)
Synonym(s):
Basic Brown 1, Basic Brown G, GX, GXP, Excelsior Brown, Leather Brown, Manchester Brown, Phenylene Brown, Vesuvine
Empirical Formula (Hill Notation):
C18H18N8 · 2HCl
CAS Number:
Molecular Weight:
419.31
EC Number:

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing