Skip to Content
Merck
Bis(cyclopentadienyl)magnesium(II) purified by sublimation, 97%

Bis(cyclopentadienyl)magnesium(II)

Bis(cyclopentadienyl)magnesium(II) purified by sublimation, 97%
Linear Formula:
Mg(C5H5)2
CAS Number:
Molecular Weight:
154.49

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing