
Bis(4-methoxyphenyl)chlorophosphine

Synonym(s):
P,P-Bis(4-methoxyphenyl)phosphinous chloride
Empirical Formula (Hill Notation):
C14H14ClO2P
CAS Number:
Molecular Weight:
280.69
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing