Skip to Content
Merck
Bis[3-(trimethoxysilyl)propyl]amine technical grade, ≥90%

Bis[3-(trimethoxysilyl)propyl]amine

Bis[3-(trimethoxysilyl)propyl]amine technical grade, ≥90%
Synonym(s):
3,3′-Bis(trimethoxysilyl)dipropylamine
Linear Formula:
[(CH3O)3Si(CH2)3]2NH
CAS Number:
Molecular Weight:
341.55
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing