Skip to Content
Merck
Benzyltrimethylammonium tetrachloroiodate ≥98.0% (AT)

Benzyltrimethylammonium tetrachloroiodate

Benzyltrimethylammonium tetrachloroiodate ≥98.0% (AT)
Linear Formula:
C6H5CH2N(ICl4)(CH3)3
CAS Number:
Molecular Weight:
418.96

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing