
Benzo(k)fluoranthene solution

Synonym(s):
Benzo[k]fluoranthene solution
Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Products
Brand
Formula Weight
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing