Skip to Content
Merck
β-Alanine methyl ester hydrochloride ≥98.0% (AT)

β-Alanine methyl ester hydrochloride

β-Alanine methyl ester hydrochloride ≥98.0% (AT)
Synonym(s):
Methyl 3-aminopropionate hydrochloride
Linear Formula:
NH2CH2CH2COOCH3 · HCl
CAS Number:
Molecular Weight:
139.58

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing