Skip to Content
Merck
α-Methylferrocenemethanol 97%

α-Methylferrocenemethanol

α-Methylferrocenemethanol 97%
Synonym(s):
1-(Ferrocenyl)ethanol, α-Ferrocenylethanol, α-Methylferrocenemethanol, (1-Hydroxyethyl)ferrocene, NSC 162110
Empirical Formula (Hill Notation):
C12H14FeO
CAS Number:
Molecular Weight:
230.08

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing