Skip to Content
Merck
Allyl P,P-diethylphosphonoacetate 97%

Allyl P,P-diethylphosphonoacetate

Allyl P,P-diethylphosphonoacetate 97%
Synonym(s):
Phosphonoacetic acid P,P-diethyl allyl ester
Linear Formula:
(C2H5O)2P(O)CH2COOCH2CH=CH2
CAS Number:
Molecular Weight:
236.20

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing