Skip to Content
Merck
Alisol B acetate ≥98% (HPLC)

Alisol B acetate

Alisol B acetate ≥98% (HPLC)
Synonym(s):
23-Acetylalismol B, 23-O-Acetylalisol B, Alisol B 23-acetate
Empirical Formula (Hill Notation):
C32H50O5
CAS Number:
Molecular Weight:
514.74

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing