Skip to Content
Merck
9-Chloro-9-phenylxanthene 97%

9-Chloro-9-phenylxanthene

9-Chloro-9-phenylxanthene 97%
Synonym(s):
Pixyl chloride
Empirical Formula (Hill Notation):
C19H13ClO
CAS Number:
Molecular Weight:
292.76
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing