
9-Anthraceneboronic acid

Synonym(s):
9-Anthracenylboronic acid, 9-Anthrylboronic acid
Empirical Formula (Hill Notation):
C14H11BO2
CAS Number:
Molecular Weight:
222.05
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing