Skip to Content
Merck
8-(Tosylamino)quinoline 97%

8-(Tosylamino)quinoline

8-(Tosylamino)quinoline 97%
Synonym(s):
N-(8-Quinolyl)-p-toluenesulfonamide
Empirical Formula (Hill Notation):
C16H14N2O2S
CAS Number:
Molecular Weight:
298.36

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing