
8-Quinolinethiol hydrochloride

Synonym(s):
8-Mercaptoquinoline hydrochloride, Thiooxine hydrochloride
Empirical Formula (Hill Notation):
C9H7NS · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing