Skip to Content
Merck
7-Hydroxy-4-(trifluoromethyl)coumarin 98%

7-Hydroxy-4-(trifluoromethyl)coumarin

7-Hydroxy-4-(trifluoromethyl)coumarin 98%
Synonym(s):
4-(Trifluoromethyl)umbelliferone
Empirical Formula (Hill Notation):
C10H5F3O3
CAS Number:
Molecular Weight:
230.14

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing