Skip to Content
Merck
7-Chloroquinaldine 99%

7-Chloroquinaldine

7-Chloroquinaldine 99%
Synonym(s):
7-Chloro-2-methylquinoline
Empirical Formula (Hill Notation):
C10H8ClN
CAS Number:
Molecular Weight:
177.63

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing