Skip to Content
Merck
7-bromo-1-hydroxyisoquinoline 95%

7-bromo-1-hydroxyisoquinoline

7-bromo-1-hydroxyisoquinoline 95%
Empirical Formula (Hill Notation):
C9H6BrNO
CAS Number:
Molecular Weight:
224.05

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing