Skip to Content
Merck
7-Bromo-1-chloro-isoquinoline 97%

7-Bromo-1-chloro-isoquinoline

7-Bromo-1-chloro-isoquinoline 97%
Empirical Formula (Hill Notation):
C9H5BrClN
Molecular Weight:
242.50

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing