![6-(Trifluoromethyl)uracil 97%](/deepweb/assets/sigmaaldrich/product/structures/606/400/de1d5997-e3db-4a75-91a0-9d8a3c55cf14/640/de1d5997-e3db-4a75-91a0-9d8a3c55cf14.png)
6-(Trifluoromethyl)uracil
![6-(Trifluoromethyl)uracil 97%](/deepweb/assets/sigmaaldrich/product/structures/606/400/de1d5997-e3db-4a75-91a0-9d8a3c55cf14/640/de1d5997-e3db-4a75-91a0-9d8a3c55cf14.png)
Empirical Formula (Hill Notation):
C5H3F3N2O2
CAS Number:
Molecular Weight:
180.08
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing