Skip to Content
Merck
6-Methylpyridine-3-carboxylic acid 99%

6-Methylpyridine-3-carboxylic acid

6-Methylpyridine-3-carboxylic acid 99%
Synonym(s):
6-Methylnicotinic acid
Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing