Skip to Content
Merck
6-Methoxy-1-indanone ≥97%

6-Methoxy-1-indanone

6-Methoxy-1-indanone ≥97%
Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing