Skip to Content
Merck
6-Methoxy-1,3-benzodioxole-5-carbaldehyde 97%

6-Methoxy-1,3-benzodioxole-5-carbaldehyde

6-Methoxy-1,3-benzodioxole-5-carbaldehyde 97%
Synonym(s):
5-Formyl-6-methoxy-1,3-benzodioxole, 6-Methoxypiperonal
Empirical Formula (Hill Notation):
C9H8O4
CAS Number:
Molecular Weight:
180.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing