Skip to Content
Merck
6-Hydroxy-3,4-dihydro-2(1H)-quinolinone 97%

6-Hydroxy-3,4-dihydro-2(1H)-quinolinone

6-Hydroxy-3,4-dihydro-2(1H)-quinolinone 97%
Synonym(s):
6-Hydroxy-1,2,3,4-tetrahydro-2-quinolinone, 6-Hydroxy-3,4-dihydrocarbstyril
Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing