![6-Bromoimidazo[1,2-a]pyrimidine 97%](/deepweb/assets/sigmaaldrich/product/structures/203/733/3a92231d-ec95-4397-a9fc-10e84132f578/640/3a92231d-ec95-4397-a9fc-10e84132f578.png)
6-Bromoimidazo[1,2-a]pyrimidine
![6-Bromoimidazo[1,2-a]pyrimidine 97%](/deepweb/assets/sigmaaldrich/product/structures/203/733/3a92231d-ec95-4397-a9fc-10e84132f578/640/3a92231d-ec95-4397-a9fc-10e84132f578.png)
Empirical Formula (Hill Notation):
C6H4BrN3
CAS Number:
Molecular Weight:
198.02
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing