Skip to Content
Merck
6,7-Dihydro-4-benzo[b]thiophenone 98%

6,7-Dihydro-4-benzo[b]thiophenone

6,7-Dihydro-4-benzo[b]thiophenone 98%
Synonym(s):
4-Keto-4,5,6,7-tetrahydrothianaphthene, 4-Oxo-4,5,6,7-tetrahydrobenzo[b]thiophene, 4-Oxo-4,5,6,7-tetrahydrothianaphthene
Empirical Formula (Hill Notation):
C8H8OS
CAS Number:
Molecular Weight:
152.21
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing