Skip to Content
Merck
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione 98%

6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione

6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione 98%
Synonym(s):
2,2-Dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedione
Linear Formula:
CF3CF2CF2COCH2COC(CH3)3
CAS Number:
Molecular Weight:
296.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing