
5-Nitrobarbituric acid hydrate

Synonym(s):
5-Nitro-2,4,6-trihydroxypyrimidine, Dilituric Acid
Empirical Formula (Hill Notation):
C4H3N3O5 · xH2O
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing