Skip to Content
Merck
5-Methylpyrimidine 96%

5-Methylpyrimidine

5-Methylpyrimidine 96%
Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing