Skip to Content
Merck
5-Chloro-2-thienylboronic acid

5-Chloro-2-thienylboronic acid

5-Chloro-2-thienylboronic acid
Synonym(s):
(5-Chlorothiophen-2-yl)boronic acid, 5-Chlorothiophene-2-boronic acid
Empirical Formula (Hill Notation):
C4H4BClO2S
CAS Number:
Molecular Weight:
162.40

Products

Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing