Skip to Content
Merck
5-Carboxyfluorescein diacetate ~95% (HPLC)

5-Carboxyfluorescein diacetate

5-Carboxyfluorescein diacetate ~95% (HPLC)
Synonym(s):
5-CFDA, 5-Carboxy-di-O-acetylfluorescein
Empirical Formula (Hill Notation):
C25H16O9
CAS Number:
Molecular Weight:
460.39

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing