Skip to Content
Merck
5-Bromothiophene-3-carboxylic acid 97%

5-Bromothiophene-3-carboxylic acid

5-Bromothiophene-3-carboxylic acid 97%
Empirical Formula (Hill Notation):
C5H3BrO2S
CAS Number:
Molecular Weight:
207.05

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing