
5-Bromo-2-(trifluoromethyl)pyridine

Empirical Formula (Hill Notation):
C6H3BrF3N
CAS Number:
Molecular Weight:
225.99
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing