
5-Benzyloxyindole-3-acetic acid

Synonym(s):
NSC 68361
Empirical Formula (Hill Notation):
C17H15NO3
CAS Number:
Molecular Weight:
281.31
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing