Skip to Content
Merck
5-Aminoindole hydrochloride 98%

5-Aminoindole hydrochloride

5-Aminoindole hydrochloride 98%
Synonym(s):
5-Indolamine hydrochloride
Empirical Formula (Hill Notation):
C8H8N2 · HCl
CAS Number:
Molecular Weight:
168.62

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing