Skip to Content
Merck
5,6-Dichloropyridine-3-carboxylic acid 96%

5,6-Dichloropyridine-3-carboxylic acid

5,6-Dichloropyridine-3-carboxylic acid 96%
Synonym(s):
5,6-Dichloronicotinic acid, 5,6-Dichloropyridine-3-carboxylic acid
Empirical Formula (Hill Notation):
C6H3Cl2NO2
CAS Number:
Molecular Weight:
192.00

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing