
5,10,15,20-Tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate)

Synonym(s):
meso-Tetra(N-methyl-4-pyridyl)porphine tetratosylate salt, TMPyP
Empirical Formula (Hill Notation):
C72H66N8O12S4
CAS Number:
Molecular Weight:
1363.60
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing