Skip to Content
Merck
4-(Trimethylsiloxy)-3-penten-2-one 97%

4-(Trimethylsiloxy)-3-penten-2-one

4-(Trimethylsiloxy)-3-penten-2-one 97%
Synonym(s):
Acetylacetone enol trimethylsilyl ether, TMS acac
Linear Formula:
(CH3)3SiOC(CH3)=CHCOCH3
CAS Number:
Molecular Weight:
172.30
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing