4-tert-Octylphenol-3,5-d2 diethoxylate solution
Synonym(s):
2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxyl-3,5-d2]ethoxy}ethanol, 4-tert-OP2EO-D2
Empirical Formula (Hill Notation):
C18D2H28O3
Products
Brand
Formula Weight
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing