Skip to Content
Merck
4-Sulfophenyl isothiocyanate sodium salt monohydrate technical grade

4-Sulfophenyl isothiocyanate sodium salt monohydrate

4-Sulfophenyl isothiocyanate sodium salt monohydrate technical grade
Synonym(s):
4-Isothiocyanatobenzenesulfonic acid sodium salt monohydrate
Linear Formula:
NaO3SC6H4NCS·H2O
CAS Number:
Molecular Weight:
255.25
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing