Skip to Content
Merck
4-Pyridazinecarboxylic acid 97%

4-Pyridazinecarboxylic acid

4-Pyridazinecarboxylic acid 97%
Empirical Formula (Hill Notation):
C5H4N2O2
CAS Number:
Molecular Weight:
124.10

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing