Skip to Content
Merck
4′-(Methylsulfonyl)acetophenone 97%

4′-(Methylsulfonyl)acetophenone

4′-(Methylsulfonyl)acetophenone 97%
Synonym(s):
1-[4-(Methylsulfonyl)phenyl]ethan-1-one, 4-(Methylsulfonyl)acetophenone, NSC 403928
Linear Formula:
CH3SO2C6H4COCH3
CAS Number:
Molecular Weight:
198.24
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing