Skip to Content
Merck
4-Methyl-2-pyridinecarbonitrile 97%

4-Methyl-2-pyridinecarbonitrile

4-Methyl-2-pyridinecarbonitrile 97%
Synonym(s):
2-Cyano-4-methylpyridine, 4-Methyl-2-cyanopyridine
Empirical Formula (Hill Notation):
C7H6N2
CAS Number:
Molecular Weight:
118.14

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing