Skip to Content
Merck
4-Methoxypyridin-2-ylamine 95%

4-Methoxypyridin-2-ylamine

4-Methoxypyridin-2-ylamine 95%
Synonym(s):
4-Methoxy-pyridin-2-ylamine
Empirical Formula (Hill Notation):
C6H8N2O
CAS Number:
Molecular Weight:
124.14

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing