Skip to Content
Merck
4′-Hydroxy-3′-nitroacetophenone 98%

4′-Hydroxy-3′-nitroacetophenone

4′-Hydroxy-3′-nitroacetophenone 98%
Linear Formula:
HOC6H3(NO2)COCH3
CAS Number:
Molecular Weight:
181.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing