4′-Hydroxy-2,4-dimethoxybenzophenone
Synonym(s):
(2,4-Dimethoxyphenyl)-(4-hydroxyphenyl)methanone, 2,4-Dimethoxy-4′-hydroxybenzophenone
Linear Formula:
(CH3O)2C6H3COC6H4OH
CAS Number:
Molecular Weight:
258.27
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing