Skip to Content
Merck
4-Hydrazonomethyl-1-hydroxy-2,2,5,5-tetramethyl-3-imidazoline-3-oxide ≥95.0% (NT)

4-Hydrazonomethyl-1-hydroxy-2,2,5,5-tetramethyl-3-imidazoline-3-oxide

4-Hydrazonomethyl-1-hydroxy-2,2,5,5-tetramethyl-3-imidazoline-3-oxide ≥95.0% (NT)
Synonym(s):
1-Hydroxy-2,2,5,5-tetramethyl-3-imidazoline-4-carboxaldehyde hydrazone-3-oxide, 2,5-Dihydro-1-hydroxy-2,2,5,5-tetramethylimidazole-4-carboxaldehyde hydrazone 3-oxide, HHTI
Empirical Formula (Hill Notation):
C8H16N4O2
CAS Number:
Molecular Weight:
200.24

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1