Skip to Content
Merck
4-Guanidinobenzoic acid hydrochloride 99%

4-Guanidinobenzoic acid hydrochloride

4-Guanidinobenzoic acid hydrochloride 99%
Synonym(s):
4-Guanidobenzoic acid hydrochloride
Linear Formula:
H2NC(=NH)NHC6H4CO2H · HCl
CAS Number:
Molecular Weight:
215.64
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing